Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006691.3 | NP_006682.2 | 6 | 1740 | ||
GenBank mRNA | AF118108.1 | - | 6 | 1544 | ||
AY358925.1 | - | 6 | 1613 | |||
BC026231.1 | - | 6 | 1499 | 4712863 |