Search
Search

FAM219B| Gene Symbol: FAM219B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321922.1 | NP_001308851.1 | 5 | 1061 | ||
| NM_020447.4 | NP_065180.1 | 5 | 1064 | |||
| NR_135841.1 | 5 | 1034 | ||||
| NR_135842.1 | 5 | 1054 | ||||
| NR_135843.1 | 4 | 973 | ||||
| NR_135845.1 | 6 | 1142 | ||||
| NR_135848.1 | 4 | 976 | ||||
| NR_135849.1 | 6 | 1122 | ||||
| NR_135850.1 | 5 | 1051 | ||||
| NR_135851.1 | 6 | 1139 | ||||
| NR_135854.1 | 5 | 1031 | ||||
| GenBank mRNA | AK000005.1 | - | 3 | 2453 | ||
| AK001572.1 | - | 4 | 962 | |||
| AK291020.1 | - | 5 | 1051 | |||
| AK296403.1 | - | 6 | 1129 | |||
| AK308954.1 | - | 4 | 960 | |||
| BC015637.2 | - | 5 | 1008 | 4636850 | ||