Search
Search

Gm10590
4933409K07Rik
Gm7819| Gene Symbol: Gm10590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048672.1 | - | 1313 | |||
| Gene Symbol: 4933409K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072647.1 | - | 5 | 1393 | 30532552 | |
| Gene Symbol: Gm7819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029949.1 | - | 5 | 1210 | ||