Search
Search

IL1F10| Gene Symbol: IL1F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032556.5 | NP_115945.4 | 4 | 1060 | ||
| NM_173161.2 | NP_775184.1 | 5 | 714 | |||
| GenBank mRNA | AF334755.1 | - | 4 | 1060 | ||
| AY026753.1 | - | 2 | 705 | |||
| AY029413.1 | - | 5 | 714 | |||
| AY358846.1 | - | 5 | 702 | |||