Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006691.3 | NP_006682.2 | 2 | 393 | ||
GenBank mRNA | AF118108.1 | - | 2 | 197 | ||
AF127670.2 | - | 2 | 227 | |||
AK075443.1 | - | 2 | 307 | |||
AK312462.1 | - | 2 | 391 | |||
AY358925.1 | - | 2 | 266 | |||
BC026231.1 | - | 2 | 152 | 4712863 | ||
DA924658.1 | - | 2 | 393 |