Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGDIA SIRNA Assays ›
Gene Symbol:
ARHGDIA
Gene Name:
Rho GDP dissociation inhibitor alpha
Gene Aliases:
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
Chromosome Location:
Chr.17: 81867719 - 81871406 on Build GRCh38
UniGene ID:
Hs.159161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46085
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGDIA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001185077.2
NP_001172006.1
2
440
NM_001185078.2
NP_001172007.1
2
210
NM_001301240.1
NP_001288169.1
2
210
NM_001301241.1
NP_001288170.1
2
440
NM_001301242.1
NP_001288171.1
2
210
NM_001301243.1
NP_001288172.1
2
210
NM_004309.5
NP_004300.1
2
210
NR_125441.1
2
210
XM_011523574.1
XP_011521876.1
2
435
GenBank mRNA
AF498926.1
-
1
74
AK293147.1
-
2
154
AK300816.1
-
2
183
AK312347.1
-
2
146
BC005851.2
-
2
166
2961556
BC005875.2
-
2
166
2961556
BC008701.3
-
1
101
2966760
BC009759.2
-
2
131
3935956
BC013846.1
-
2
130
BC016031.1
-
1
82
3911525
BC016185.1
-
2
126
4867857
BC024258.1
-
2
126
3027721
BC027730.2
-
2
129
4764249
BC028333.1
-
2
129
BC075827.1
-
2
126
6074977
BC106044.1
-
2
169
6162284
BI755343.1
-
2
131
BI768191.1
-
2
162
BM552638.1
-
2
380
BP325393.1
-
2
141
BQ674863.1
-
2
124
BT006884.1
-
1
74
CF132249.1
-
2
134
CF132324.1
-
2
440
CR456777.1
-
1
74
D13989.1
-
1
94
DA433004.1
-
2
129
DC406556.1
-
2
210
EU794615.1
-
1
101
KU177916.1
-
1
74
KU177917.1
-
1
74
M97579.1
-
1
74
X69550.1
-
2
127
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
negative regulation of cell adhesion
regulation of Rho protein signal transduction
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of axonogenesis
positive regulation of axonogenesis
regulation of small GTPase mediated signal transduction
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0