Search
Search

S100g| Gene Symbol: S100g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012521.2 | NP_036653.1 | 2 | 133 | ||
| XM_006256888.2 | XP_006256950.1 | 2 | 508 | |||
| GenBank mRNA | BC059153.1 | - | 2 | 137 | 6917928 | |
| J02954.1 | - | 2 | 134 | |||