Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033675.1 | NP_001028847.1 | 5 | 434 | ||
NM_001033676.1 | NP_001028848.1 | 5 | 1046 | |||
NM_133529.2 | NP_598213.1 | 6 | 614 | |||
GenBank mRNA | AJ315657.1 | - | 6 | 614 | ||
AJ315761.1 | - | 5 | 434 | |||
Y17048.2 | - | 5 | 1046 |