Search
Search

SHH| Gene Symbol: SHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_000193.3 | NP_000184.1 | 2 | 677 | ||
| NM_001310462.1 | NP_001297391.1 | 3 | 435 | |||
| NR_132318.1 | 3 | 435 | ||||
| NR_132319.1 | 3 | 435 | ||||
| XM_011516479.2 | XP_011514781.1 | 3 | 3177 | |||
| XM_011516480.2 | XP_011514782.1 | 4 | 3475 | |||
| GenBank mRNA | AY927450.1 | - | LIT3478 AY927450 |
3 | 427 | |
| AY927452.1 | - | LIT3480 AY927452 |
3 | 435 | ||
| AY927453.1 | - | 3 | 435 | |||
| AY927454.1 | - | 3 | 435 | |||
| AY927455.1 | - | LIT3481 AY927455 |
3 | 435 | ||
| BX452210.1 | - | 2 | 582 | |||
| L38518.1 | - | 2 | 677 | |||