Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003463.4 | NP_003454.1 | 2 | 1236 | ||
XM_011536111.1 | XP_011534413.1 | 2 | 575 | |||
XM_011536112.1 | XP_011534414.1 | 2 | 1334 | |||
XM_017011270.1 | XP_016866759.1 | 2 | 592 | |||
XM_017011271.1 | XP_016866760.1 | 2 | 572 | |||
XM_017011272.1 | XP_016866761.1 | 2 | 1236 | |||
GenBank mRNA | AB451314.1 | - | 1 | 83 | ||
AB451444.1 | - | 1 | 83 | |||
AK312526.1 | - | 2 | 720 | |||
BC023975.2 | - | 2 | 678 | 2960001 | ||
BC045571.1 | - | 2 | 1236 | 4826233 | ||
U48296.1 | - | 1 | 475 |