Search
Search

PMFBP1| Gene Symbol: PMFBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160213.1 | NP_001153685.1 | 7 | 1104 | ||
| NM_031293.2 | NP_112583.2 | 6 | 926 | |||
| XM_011523357.2 | XP_011521659.1 | 6 | 1016 | |||
| XM_011523358.2 | XP_011521660.1 | 6 | 956 | |||
| XM_011523360.2 | XP_011521662.1 | 6 | 979 | |||
| XM_011523361.2 | XP_011521663.1 | 6 | 1454 | |||
| XM_011523362.2 | XP_011521664.1 | 5 | 761 | |||
| GenBank mRNA | AK093285.1 | - | 6 | 922 | ||
| AK097465.1 | - | 2 | 163 | |||
| AK122925.1 | - | 7 | 1104 | |||
| AK315004.1 | - | 6 | 907 | |||
| AL136795.1 | - | 6 | 886 | |||
| BC028423.2 | - | 6 | 913 | 4822328 | ||