Search
Search

ZMYM6| Gene Symbol: ZMYM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007167.3 | NP_009098.3 | 6 | 944 | ||
| GenBank mRNA | AK299293.1 | - | 6 | 903 | ||
| AK301461.1 | - | 5 | 712 | |||
| AK310703.1 | - | 6 | 885 | |||
| AK310938.1 | - | 4 | 444 | |||
| AL389985.1 | - | 6 | 869 | |||
| AL833263.1 | - | 5 | 667 | |||
| BC033903.1 | - | 6 | 926 | |||
| BC107880.1 | - | 6 | 925 | |||