Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS146271
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
112
20510
NM_182910.2
NP_878914.1
6
1283
NM_182913.2
NP_878917.1
8
1388
NM_182914.2
NP_878918.2
113
20579
XM_005267454.1
XP_005267511.1
113
20621
XM_005267456.1
XP_005267513.1
113
20621
XM_005267457.1
XP_005267514.1
113
20579
XM_005267458.1
XP_005267515.1
112
20552
XM_005267459.1
XP_005267516.1
112
20510
XM_011536574.1
XP_011534876.1
113
20663
XM_011536575.2
XP_011534877.1
113
20640
XM_011536576.2
XP_011534878.1
114
20820
XM_011536577.2
XP_011534879.1
113
20587
XM_011536578.1
XP_011534880.1
113
20663
XM_011536579.1
XP_011534881.1
113
20621
XM_011536580.1
XP_011534882.1
113
20621
XM_011536581.1
XP_011534883.1
112
20594
XM_011536582.1
XP_011534884.1
112
20546
XM_017021101.1
XP_016876590.1
113
20665
XM_017021102.1
XP_016876591.1
112
20708
XM_017021103.1
XP_016876592.1
15
2700
XM_017021104.1
XP_016876593.1
15
2700
GenBank mRNA
AB023228.3
-
64
10251
AF357236.1
-
112
20049
AF435011.1
-
112
20492
AF495911.1
-
112
20477
AK001876.1
-
3
422
AK074055.1
-
24
3977
AK090430.1
-
30
4886
AK297874.1
-
8
1307
AK298465.1
-
7
1021
AL080133.1
-
18
3023
AL117404.1
-
6
1805
AY061757.1
-
13
2114
AY061758.1
-
8
1385
AY061759.1
-
60
9813
AY184204.1
-
6
1805
AY184205.1
-
11
1995
BX537642.1
-
33
5324
BX538095.1
-
21
3486
BX640974.1
-
14
2325
CR749324.1
-
33
5484
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0