Search
Search

MGAM2| Gene Symbol: MGAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293626.1 | NP_001280555.1 | 11 | 1217 | ||
| XM_011516692.2 | XP_011514994.1 | 4 | 432 | |||
| GenBank mRNA | AK000645.1 | - | 11 | 1217 | ||
| BC111973.1 | - | 11 | 1224 | |||
| BC111975.1 | - | 11 | 1224 | |||