Search
Search

NUDT7| Gene Symbol: NUDT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105663.2 | NP_001099133.1 | 3 | 307 | ||
| NM_001243657.1 | NP_001230586.1 | 3 | 307 | |||
| XM_011523032.2 | XP_011521334.1 | 4 | 425 | |||
| XM_011523033.2 | XP_011521335.1 | 3 | 311 | |||
| XM_017023170.1 | XP_016878659.1 | 4 | 425 | |||
| GenBank mRNA | AA227330.1 | - | 3 | 259 | ||
| BU579534.1 | - | 3 | 253 | |||
| BX494638.1 | - | 3 | 286 | |||
| CN297379.1 | - | 3 | 258 | |||
| ES309123.1 | - | 1 | 42 | |||