Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015030.1 | NP_055845.1 | 55, 56 | 8292 | ||
XM_005248090.4 | XP_005248147.1 | 56, 57 | 8350 | |||
XM_005248093.4 | XP_005248150.1 | 30, 31 | 4977 | |||
XM_011513680.2 | XP_011511982.1 | 53, 54 | 7809 | |||
XM_011513681.2 | XP_011511983.1 | 53, 54 | 7812 | |||
XM_011513682.2 | XP_011511984.1 | 52, 53 | 7767 | |||
XM_011513683.2 | XP_011511985.1 | 55, 56 | 8186 | |||
XM_011513684.2 | XP_011511986.1 | 30, 31 | 4983 | |||
XM_017008046.1 | XP_016863535.1 | 56, 57 | 8617 | |||
GenBank mRNA | AB020633.1 | - | 13, 14 | 2356 | ||
AK127307.1 | - | 17, 18 | 3037 | |||
AL833170.1 | - | 22, 23 | 3813 |