Search
Search

C21orf2| Gene Symbol: C21orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271440.1 | NP_001258369.1 | 2, 3 | 310 | ||
| NM_001271441.1 | NP_001258370.1 | 2, 3 | 310 | |||
| NM_004928.2 | NP_004919.1 | 2, 3 | 310 | |||
| XM_017028470.1 | XP_016883959.1 | 2, 3 | 321 | |||
| GenBank mRNA | AB209578.1 | - | 2, 3 | 576 | ||
| BC031300.1 | - | 2, 3 | 301 | 4878257 | ||
| BC072012.1 | - | 2, 3 | 283 | 6645626 | ||
| BM761336.1 | - | 2, 3 | 310 | |||
| U84569.1 | - | 2, 3 | 282 | |||
| Y11392.1 | - | 2, 3 | 170 | |||
| Z93322.1 | - | 2, 3 | 109 | |||