Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 4 | 391 | ||
NM_011431.3 | NP_035561.1 | 4 | 388 | |||
GenBank mRNA | AK052242.1 | - | 4 | 346 | ||
AK150158.1 | - | 4 | 360 | |||
AK153473.1 | - | 4 | 360 | |||
AK154878.1 | - | 4 | 370 | |||
AK165151.1 | - | 4 | 378 | |||
AK165624.1 | - | 4 | 362 | |||
AK172875.1 | - | 4 | 342 | |||
AK190907.1 | - | 2 | 148 | |||
AK193531.1 | - | 2 | 148 | |||
AK195071.1 | - | 2 | 148 | |||
AK196741.1 | - | 2 | 175 | |||
AK202497.1 | - | 2 | 148 | |||
AK203192.1 | - | 2 | 148 | |||
AK203588.1 | - | 2 | 149 | |||
AK219617.1 | - | 2 | 70 | |||
BC052674.1 | - | 4 | 364 | 30061580 | ||
BC054778.1 | - | 4 | 354 | 6810127 | ||
BQ886207.1 | - | 4 | 383 | |||
U97079.1 | - | 4 | 330 |