Search
Search

Bcas1| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164369.1 | NP_001157841.1 | 9 | 1324 | ||
| NM_029815.2 | NP_084091.2 | 10 | 1462 | |||
| GenBank mRNA | AK008957.1 | - | 7 | 868 | ||
| AY219233.1 | - | 10 | 1445 | |||
| BC113166.1 | - | 9 | 1304 | 40059981 | ||
| BC113957.1 | - | 9 | 1304 | 40059979 | ||