Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027935.2 | NP_082211.1 | 2 | 54 | ||
GenBank mRNA | AK014282.1 | - | 2 | 124 | ||
AK147069.1 | - | 2 | 90 | |||
AK152630.1 | - | 2 | 138 | |||
AK152654.1 | - | 2 | 109 | |||
AK157214.1 | - | 2 | 127 | |||
AK164989.1 | - | 2 | 120 | |||
AK165636.1 | - | 2 | 103 | |||
AK179839.1 | - | 1 | 103 | |||
BC094256.1 | - | 2 | 54 | 6820666 |