Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700123I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165919.1 | NP_001159391.1 | 10, 11 | 914 | ||
GenBank mRNA | AK007250.1 | - | 5, 6 | 524 | ||
BC138005.1 | - | 10, 11 | 828 | 8861024 | ||
BC145092.1 | - | 10, 11 | 828 | 9053640 |