Search
Search

Hoxc9| Gene Symbol: Hoxc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008272.3 | NP_032298.1 | 2 | 855 | ||
| GenBank mRNA | AK012173.1 | - | 1 | 243 | ||
| AK078758.1 | - | 1 | 281 | |||
| BC050838.1 | - | 2 | 852 | 6510837 | ||
| X55318.1 | - | 1 | 572 | |||