Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001985.3 | NP_001001985.3 | 2 | 1369 | ||
GenBank mRNA | AK147307.1 | - | 2 | 1370 | ||
AK147594.1 | - | 2 | 648 | |||
AK147703.1 | - | 2 | 707 | |||
BC075616.1 | - | 2 | 450 | |||
DQ174094.1 | - | 2 | 450 |