Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029985.4 | NP_001025156.3 | 17, 18 | 1844 | ||
GenBank mRNA | AK154369.1 | - | 17, 18 | 1832 | ||
AY884211.1 | - | 17, 18 | 1830 | |||
BQ179923.1 | - | 1 | 14 |