Search
Search

Zfp180| Gene Symbol: Zfp180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271280.1 | NP_001258209.1 | 5 | 849 | ||
| NM_144757.3 | NP_653358.2 | 5 | 767 | |||
| XM_006228386.2 | XP_006228448.1 | 5 | 800 | |||
| XM_006228387.2 | XP_006228449.1 | 5 | 851 | |||
| XM_008758890.1 | XP_008757112.1 | 319 | ||||
| GenBank mRNA | U41164.1 | - | 5 | 767 | ||