Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030001.2 | NP_001025172.1 | 2 | 208 | ||
NM_001032.4 | NP_001023.1 | 2 | 208 | |||
GenBank mRNA | AK026649.1 | - | 1 | 168 | ||
AK307833.1 | - | 2 | 169 | |||
BC015974.1 | - | 2 | 172 | 4105424 | ||
BC032813.1 | - | 1 | 143 | 5217083 | ||
BC035313.1 | - | 143 | 3868721 | |||
BF128896.1 | - | 2 | 115 | |||
BQ433530.1 | - | 2 | 152 | |||
BU942244.1 | - | 2 | 211 | |||
L31610.1 | - | 2 | 149 | |||
U14973.1 | - | 2 | 168 |