Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLOT1 SIRNA Assays ›
          Gene Symbol:
          FLOT1
          Gene Name:
          flotillin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 30727709 - 30742851 on Build GRCh38
          UniGene ID:
          Hs.179986
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19915

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FLOT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318875.1 NP_001305804.1 7 841
          NM_005803.3 NP_005794.1 8 985
          XM_005248780.3 XP_005248837.1 8 718
          XM_006714947.3 XP_006715010.1 7 822
          XM_017010157.1 XP_016865646.1 8 639
          XM_017010158.1 XP_016865647.1 6 540
          GenBank mRNA AF085357.1 - 8 774
          AF089750.2 - 8 810
          AF117234.1 - 8 811
          AK000816.1 - 8 821
          AK222530.1 - 8 821
          AK301291.1 - 7 675
          BC001146.1 - 7 668 3534771
          BC128154.1 - 1 78
          BT007077.1 - 7 646
          CR456946.1 - 7 646

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          membrane raft assembly
          positive regulation of cytokine production
          positive regulation of protein phosphorylation
          regulation of receptor internalization
          axonogenesis
          extracellular matrix disassembly
          positive regulation of protein binding
          positive regulation of synaptic transmission, dopaminergic
          positive regulation of interferon-beta production
          dsRNA transport
          positive regulation of heterotypic cell-cell adhesion
          positive regulation of toll-like receptor 3 signaling pathway
          regulation of toll-like receptor 4 signaling pathway
          response to endoplasmic reticulum stress
          regulation of Rho protein signal transduction
          plasma membrane raft assembly
          positive regulation of endocytosis
          positive regulation of skeletal muscle tissue development
          protein homooligomerization
          positive regulation of cell adhesion molecule production
          protein kinase C signaling
          cellular response to exogenous dsRNA
          establishment of protein localization to plasma membrane
          positive regulation of myoblast fusion
          positive regulation of cell junction assembly
          protein localization to membrane raft
          positive regulation of cell-cell adhesion mediated by cadherin
          protease binding
          protein binding
          ionotropic glutamate receptor binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-66c8j:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0