Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163159.1 | NP_001156631.1 | 7, 8 | 992 | ||
NM_001163160.1 | NP_001156632.1 | 7, 8 | 926 | |||
NM_009981.4 | NP_034111.1 | 7, 8 | 828 | |||
GenBank mRNA | AK076050.1 | - | 7, 8 | 753 | ||
AK076830.1 | - | 7, 8 | 927 | |||
BC018313.1 | - | 6, 7 | 711 | 4162053 | ||
L28956.1 | - | 7, 8 | 797 | |||
Z12302.1 | - | 7, 8 | 787 |