Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290048.1 | NP_001276977.1 | 12 | 1771 | ||
NM_015459.4 | NP_056274.3 | 12 | 1873 | |||
XM_006718493.1 | XP_006718556.1 | 11 | 1698 | |||
GenBank mRNA | AK023383.1 | - | 12 | 1478 | ||
AK097588.1 | - | 14 | 1813 | |||
AK301910.1 | - | 12 | 1709 | |||
AL117600.1 | - | 12 | 1520 | |||
BC077727.1 | - | 12 | 1764 | 5952471 | ||
BX642720.1 | - | 1 | 113 | |||
CR936784.1 | - | 12 | 1756 |