Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291484.2 | NP_001278413.1 | 3 | 751 | ||
NM_001308398.1 | NP_001295327.1 | 3 | 751 | |||
NM_004363.5 | NP_004354.3 | 3 | 751 | |||
XM_005258413.3 | XP_005258470.2 | 3 | 720 | |||
XM_011526322.1 | XP_011524624.1 | 3 | 720 | |||
XM_017026145.1 | XP_016881634.1 | 3 | 720 | |||
XM_017026146.1 | XP_016881635.1 | 3 | 720 | |||
GenBank mRNA | AB852566.1 | - | 5 | 1138 | ||
AB852567.1 | - | 4 | 604 | |||
AK223101.1 | - | 3 | 687 | |||
AK310844.1 | - | 3 | 710 | |||
BC034671.1 | - | 3 | 699 | 5184800 | ||
CR749337.1 | - | 5 | 861 | |||
M15042.1 | - | 3 | 593 | |||
M16234.1 | - | 3 | 683 | |||
M17191.1 | - | 3 | 729 | |||
M29540.1 | - | 3 | 718 | |||
X16455.1 | - | 3 | 593 |