Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145246.4 | NP_660289.2 | 5 | 433 | ||
XM_005269515.4 | XP_005269572.1 | 6 | 530 | |||
XM_005269516.4 | XP_005269573.1 | 5 | 625 | |||
XM_011539259.2 | XP_011537561.1 | 5 | 398 | |||
XM_011539260.2 | XP_011537562.1 | 6 | 488 | |||
XM_017015656.1 | XP_016871145.1 | 5 | 398 | |||
XM_017015657.1 | XP_016871146.1 | 5 | 433 | |||
XM_017015658.1 | XP_016871147.1 | 6 | 533 | |||
XM_017015659.1 | XP_016871148.1 | 5 | 625 | |||
XM_017015660.1 | XP_016871149.1 | 6 | 490 | |||
XM_017015661.1 | XP_016871150.1 | 6 | 547 | |||
XM_017015662.1 | XP_016871151.1 | 5 | 398 | |||
GenBank mRNA | AK090955.1 | - | 5 | 433 | ||
AK308861.1 | - | 5 | 351 | |||
BM272089.1 | - | 5 | 417 | |||
BX648699.1 | - | 4 | 467 | |||
CN298116.1 | - | 5 | 412 |