Search
Search

Slc20a1| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015747.2 | NP_056562.1 | 10 | 2330 | ||
| GenBank mRNA | AK155506.1 | - | 10 | 2303 | ||
| BC015085.1 | - | 10 | 2230 | 4015575 | ||
| M73696.1 | - | 9 | 2249 | |||