Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 5 | 767 | ||
NM_019987.2 | NP_064371.2 | 5 | 695 | |||
GenBank mRNA | AF225918.1 | - | 5 | 535 | ||
AK087484.1 | - | 5 | 618 | |||
AK133537.1 | - | 5 | 634 | |||
AK154185.1 | - | 5 | 768 | |||
BC028863.1 | - | 4 | 452 | 4224269 |