Type:
Whole Allergen
Whole Allergen
Whole Allergen
Megrim
f311
Bothidae
Lepidorhombus whiffiagonis
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Megrim, also called e.g. white sole or whiff, belongs to the order Pleuronectiformes and is one of the most frequently consumed fish in Spain.
In a study in children, the clinical symptoms induced by this fish allergy were urticaria and angioedema (92.4%) in 79 patients, followed by atopic dermatitis (17.7%) and inhalation-related asthma (13.9%) (1). In an immunoblot study of specific IgE binding, Megrim bound the greatest amount compared with the related sole and witch (1).
IgE-mediated reactions
The most common symptoms in a group of megrim-allergic children were urticaria and angioedema.
Asthma related to inhalation of cooking steam was reported.
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small
Last Reviewed- April 2022