Whole Allergen
f311 Megrim
f311 Megrim Scientific Information
Type:
Whole Allergen
Display Name:
Megrim
Allergen code:
f311
Family:
Bothidae
Latin Name:
Lepidorhombus whiffiagonis
WHO/ICD-11 code:
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Clinical Relevance
Megrim, also called e.g. white sole or whiff, belongs to the order Pleuronectiformes and is one of the most frequently consumed fish in Spain.
In a study in children, the clinical symptoms induced by this fish allergy were urticaria and angioedema (92.4%) in 79 patients, followed by atopic dermatitis (17.7%) and inhalation-related asthma (13.9%) (1). In an immunoblot study of specific IgE binding, Megrim bound the greatest amount compared with the related sole and witch (1).
IgE-mediated reactions
The most common symptoms in a group of megrim-allergic children were urticaria and angioedema.
Asthma related to inhalation of cooking steam was reported.
Molecular Aspects
Cross-reactivity
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small
Compiled By
Last Reviewed-  April 2022
References
  1. Pascual, C; Martín Esteban, M; Fernández Crespo, J. Fish allergy: Evaluation of the importance of cross-reactivity. J Pediatr; 1992; 121: S29-34.