Search
Anti-Drug Antibody (ADA) Bridging ELISA Protocol |
The assessment of immunogenicity of drugs is crucial in evaluating treatment options, particularly in clinical studies involving therapeutic antibodies. The formation of anti-drug antibodies (ADAs) induced by treatment has been associated with loss of response, hypersensitivity reactions, and severe therapy-limiting side effects. Therefore, measuring ADAs is increasingly important in evaluating a patient's response to therapy.
ADA bridging ELISA represents an innovative assay format for measuring the immunogenicity of therapeutic drugs, including proteins, monoclonal antibodies, and other biologics. In this assay, the biotinylated drug is captured on streptavidin-coated plates, and anti-drug antibodies in the sample bind to the captured drug. For the detection of bivalent anti-drug antibodies, a dye or HRP-labeled drug is used. The advantages of this technique include its high sensitivity and ability to allow high-throughput sample screening. However, the specificity of bridging ELISA assays in the complex matrix of human serum may be limited due to matrix components, soluble target molecules, or drug components that can challenge the assay. Therefore, the use of high-quality assay reagents and blocking solutions is crucial to obtaining meaningful results.
Note:The bridging ELISA protocol serves as a general recommendation for using streptavidin high binding capacity coated plates for anti-drug antibody (ADA) assays. Each laboratory should customize and implement it according to their specific requirements.
Search available antibody pairs and ELISA kits Browse available ELISA reagents
Note: When using concentrated solutions (e.g., 20X TBS Tween 20 Buffer or 20X PBS Tween 20 Buffer), ensure they are diluted to 1X with sterile, distilled water. The protocol allows flexibility in using either colorimetric or chemiluminescent substrates. For colorimetric assays, use TMB substrate solution along with a stop solution. For chemiluminescent assays, a chemiluminescent substrate is required, but a stop solution is not necessary. |