DNA and RNA Quantification
Detection technologies used for DNA and RNA quantification
Nucleic acid quantification (NAQ)—the measurement of DNA or RNA concentration—is a critical step before many downstream experiments. Alongside concentration, assessing sample purity can help to ensure accurate results. Two primary optical methods are used for NAQ: UV-Vis spectrophotometry and fluorescence measurement. Each differs in sensitivity, throughput, sample volume requirements, and the type of information provided. Selecting the right method for your workflow not only improves quantification accuracy but also helps prevent downstream experimental failures, boosting overall efficiency.
Methods of quantification
Featured instruments
We offer a range of instruments to perform nucleic acid quantification using either UV-Vis spectrophotometry or fluorescence measurements.
Quantification benefits & limitations
UV-Vis and Vis spectrophotometry
Meet your analytical challenges with our complete line of ultraviolet-visible (UV-Vis) spectrophotometers. Our award-winning designs and user-friendly software help you quantify, assess purity, and more.
Fluorometers and fluorescence
Use fluorometers to quantify, detect and monitor analytes and their reactions by measuring the intensity of the fluorescent signal from dyes attached to biological molecules as well as naturally fluorescent molecules based on signature excitation (Ex) and emission (Em) wavelengths.
Instruments for photometric UV-Vis RNA/DNA quantification
Well-known Thermo Scientific instruments for UV-Vis RNA/DNA quantification include the NanoDrop Ultra Spectrophotometer and Fluorometer for convenient, single-sample microvolume analysis, the NanoDrop Eight Spectrophotometer for 8-sample microvolume analysis, and choice of the Multiskan SkyHigh Microplate Spectrophotometer or the Varioskan ALF and Varioskan LUX Multimode Reader for up to 16 microvolume samples as well as 96- and 384-well plates.
NanoDrop Ultra Microvolume UV-Vis Spectrophotometer
NanoDrop Eight Spectrophotometer
Multiskan SkyHigh Microplate Spectrophotometer
Varioskan ALF Multimode Microplate Reader
Varioskan LUX Multimode Microplate Reader
Single sample Eight samples 6- to 384-well plates, cuvettes, μDrop Plate 6- to 384-well plates,
μDrop and μDrop Duo Plates
6- to 384-well plates,
μDrop and μDrop Duo Plates
NanoDrop Ultra Microvolume UV-Vis Spectrophotometers
NanoDrop Eight Spectrophotometer
Multiskan SkyHigh Microplate Spectrophotometer
Varioskan ALF Multimode Microplate Reader
  Varioskan LUX Multimode Microplate Reader
Lower detection limit (dsDNA) Pedestal: 1.0 ng/µL Cuvette: 0.2 ng/µL 2.0 ng/µL Depends on sample format and volume used Depends on sample format and volume used Depends on sample format and volume used
Upper detection limit (dsDNA) Pedestal: 27,500 ng/µL Cuvette: 75 ng/µL 10,000 ng/µL Depends on sample format and volume used Depends on sample format and volume used Depends on sample format and volume used
Sample formats Drop (1 µL) cuvettes Drop (1 µL) 6–384 well plates µDrop Plate (2–10 µL) cuvettes (not all models) 6–384 well plates µDrop and µDrop Duo Plates (2–10 µL) 6–384 well plates µDrop and µDrop Duo Plates (2–10 µL)
Throughput One sample at a time Up to 8 One sample at a time with cuvettes
Up to 16 with µDrop plate
Up to 96 with 96-well plates
Up to 384 with 384 well plate
Up to 16 with µDrop plate
Up to 96 with 96-well plates
Up to 384 with 384 well plate
One sample at a time with cuvettes
Up to 16 with µDrop plate
Up to 96 with 96-well plates
Up to 384 with 384 well plate
Sample volume 1 µL 1 µL As little as 2 µL depending on assay format As little as 2 µL depending on assay format As little as 2 µL depending on assay format
Processing time ≤7 seconds <20 seconds for 8 samples ~1 minute for 16 microvolume samples in the µDrop Plate ~1.1 minute for 16 microvolume samples in the µDrop Plate ~1.3 minutes for 16 microvolume samples in the µDrop Plate
Spectral data Yes Yes Yes Yes Yes
Sample purity information Yes, it provides full spectral data (A260/A280 and A260/A230 ratios) as well as Acclaro Contaminant Analysis Yes, it provides full spectral data (A260/A280 and A260/A230 ratios) as well as Acclaro Contaminant Analysis Yes, it provides full spectral data (A260/A280 and A260/A230 ratios) Yes, it provides full spectral data (A260/A280 and A260/A230 ratios) Yes, it provides full spectral data (A260/A280 and A260/A230 ratios)
Contaminants detection Yes, Acclaro Sample Intelligence technology detects protein, phenol and guanidine salts and it gives true concentration of the nucleic acids Yes, Acclaro Sample Intelligence technology detects protein, phenol and guanidine salts and it gives true concentration of the nucleic acids No, some information is available, based on the purity ratios No, some information is available, based on the purity ratios No, some information is available, based on the purity ratios
Distinguish between DNA and RNA Yes, distinguishes between mammalian DNA/RNA, bacterial DNA/RNA, and plant DNA/RNA Yes, distinguishes between mammalian DNA and mammalian RNA No No No
Oligonucleotide quantification Yes, with embedded applications and custom methods Yes, with embedded applications and custom methods Yes Yes Yes
Pre-configured protocols Yes, there are user interface touch screen and custom protocols Yes, there are user interface touch screen and custom protocols Yes, there are pre-configured protocols in the interface touch screen and SkanIt software Cloud Library, free with SkanIt Software Yes, there are pre-configured protocols in the SkanIt software Cloud Library, free with SkanIt Software Yes, there are pre-configured protocols in the SkanIt software Cloud Library, free with SkanIt Software
Thermo Fisher Connect Yes No Yes No No
21 CFR Part 11 Yes, with optional software Yes, with optional software Yes, with SkanIt Software Drug Discovery Edition Yes, with SkanIt Software Drug Discovery Edition Yes, with SkanIt Software Drug Discovery Edition
Robotic automation compatibility No No Yes Yes Yes
User interface Via a touch screen user interface or via the NanoDrop Ultra PC control Software With PC software (Nanodrop Eight Operating Software) Via a touch screen user interface or the PC Software (SkanIt Software) Via PC software (SkanIt Software) Via PC software (SkanIt Software)
Instruments for fluorescence RNA/DNA quantification
High-performing instruments for performing fluorescence DNA or RNA quantification include Thermo Scientific NanoDrop Ultra FL/FL-C and Invitrogen Qubit 4 Fluorometers for single-sample measurements, Invitrogen Qubit Flex Fluorometer for medium-throughput measurements and the Thermo Scientific Varioskan ALF and Varioskan LUX Multimode Microplate Reader for 96- to 1536-well measurements.
NanoDrop Ultra Microvolume UV-Vis Fluorometer
Qubit 4 Fluorometer
Qubit Flex Fluorometer
Varioskan ALF Multimode Microplate Reader
Varioskan LUX Multimode Microplate Reader
Single sample Single sample Eight samples 6- to 384-well plates,
μDrop and μDrop Duo Plates
6- to 1536-well plates,
μDrop and μDrop Duo Plates
NanoDrop Ultra Microvolume UV-Vis Fluorometer
Qubit 4 Fluorometer
Qubit Flex Fluorometer
Varioskan ALF Multimode Microplate Reader
Varioskan LUX Multimode Microplate Reader
Lower detection limit (dsDNA) 0.05 ng/µL*** 0.005 ng/µL* 0.005 ng/µL* Depends on the Quant-iT assay kit, standard concentrations and curve fittings used** 0.005 ng/µL Depends on the Quant-iT assay kit, standard concentrations and curve fittings used** 0.005 ng/µL
Upper detection limit (dsDNA) 1,500 ng/µL**** 4000 ng/µL* 4000 ng/µL* Depends on the Quant-iT assay kit and assay conditions Depends on the Quant-iT assay kit and assay conditions
Sample formats Sample added directly to instrument Qubit assay tubes Qubit Flex tube strips 6- to 384-well plates,
μDrop and μDrop Duo Plates
6- to 1536-well plates,
μDrop and μDrop Duo Plates
Throughput 1 1 Up to 8 Dependent on assay format Dependent on assay format
Sample volume 1–10 µL 1–20 µL 1–20 µL Dependent on assay format Dependent on assay format
Processing time ≤10 seconds <3 seconds ~3 seconds for 8 samples Dependent on assay format Dependent on assay format
Sample purity and contaminants detection No, only through absorbance applications No No No No
Distinguish between DNA and RNA Yes, using fluorescent dyes that are specific for DNA or RNA Yes, using fluorescent dyes that are specific for DNA or RNA Yes, using fluorescent dyes that are specific for DNA or RNA Yes, using fluorescent dyes that are specific for DNA or RNA Yes, using fluorescent dyes that are specific for DNA or RNA
RNA quality and integrity score No Yes, when used with Qubit RNA IQ Assay Kit Yes, when used with Qubit RNA IQ Assay Kit No No
Oligonucleotide quantification No Yes, with ssDNA assay kit Yes, with ssDNA assay kit Yes, with Quant-iT OliGreen assay kit Yes, with Quant-iT OliGreen assay kit
Pre-configured protocols Yes, there are user interface touch screen and custom protocols Yes, there are user interface touch screen and custom protocols Yes, there are pre-configured protocols in the interface touch screen and SkanIt software Cloud Library, free with SkanIt Software Yes, there are pre-configured protocols in the SkanIt Software Cloud Library, free with SkanIt Software Yes, there are pre-configured protocols in the SkanIt Software Cloud Library, free with SkanIt Software
Thermo Fisher Connect Yes Yes Yes No No
21 CFR Part 11 Yes, with optional software No Yes, with optional software Yes, with SkanIt Software Drug Discovery Edition Yes, with SkanIt Software Drug Discovery Edition
Automation No No No Yes Yes
* Reported here are the lower and upper quantitation ranges measured with Qubit dsDNA High sensitivity (HS) kit (Cat. No. Q32851) Qubit dsDNA Broad Range (BR) kit (Cat. No. Q32850) respectively and using variable sample volume (1–20 µL).
** Limit of Detection (LOD) using the Quant-iT dsDNA Assay HS kit (Cat. No. Q33120) as instructed. LOD estimated according to IUPAC, based on the precision of the blanks (3*SD). Results will vary depending on the curve fittings, standard concentrations and used kits.
*** Reported here is the lower quantitation range measured with the NanoDrop Ultra dsDNA BR Fluorescence Assay and using sample volume (10 µL).
**** Reported here is the upper quantitation range measured with the NanoDrop Ultra dsDNA BR Fluorescence Assay and using sample volume (1 µL).
Fluorescence assay kits
Frequently asked questions
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity--plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
For Illumina sequencing, library molecules contain the P5 and P7 adapter sequences required for flow cell binding, cluster generation, and sequencing.  The Qubit NGS Library Quantification Assay (Cat. No. Q34250) is a fluorescence-based assay for precise molar quantification of sequencing-ready Illumina NGS libraries. The assay selectively quantifies dual-adapted DNA libraries prior to sequencing, helping support optimal cluster density and successful sequencing runs.
For Research Use Only. Not for use in diagnostic procedures.