Use 96- and 384-well Microplates for Fluorescence-based Assays with Quant-iT assays for optimal results
Quant-iT™ 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)
Invitrogen™

Quant-iT™ 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)

For easy and accurate dsDNA quantitation, the Quant-iT 1X dsDNA HS and BR Assays enable up to 1,000 fluorescence-based assays in a 96-well microplate format.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityAssayQuantitation Range
Q332321000 assays1 x dsDNA, high sensitivity0.2 to 100 ng
Q332671 kit1 x dsDNA, broad range4 to 4000 ng
Catalog number Q33232
Price (USD)
640.65
Online exclusive
696.00
Save 55.35
Each
Quantity:
1000 assays
Assay:
1 x dsDNA, high sensitivity
Quantitation Range:
0.2 to 100 ng
Price (USD)
640.65
Online exclusive
696.00
Save 55.35
Each
Make DNA quantitation easy and accurate in a ready-to-go 1X formulation with the Quant-iT 1X dsDNA HS (High-Sensitivity) and Quant-iT 1X dsDNA BR (Broad Range) Assay kits. Both kits provide a ready-to-use working solution. Simply load your sample, add the Qubit 1X dsDNA HS or BR working solution, and read the concentration using a fluorescence-based microplate reader.
The Quant-iT 1X dsDNA HS and BR Assay kits are highly selective for double-stranded DNA (dsDNA) over RNA, providing an accurate and precise method for the quantification of dsDNA samples. To perform the assay, dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a fluorescence plate reader.

The Quant-iT 1X dsDNA HS Assay Kit is accurate for initial sample concentrations ranging from 10 pg/μL to 100 ng/μL, providing a core detection range of 0.2 ng to 100 ng of DNA in the assay tube. The Quant-iT 1X dsDNA BR Assay Kit is accurate for initial DNA sample concentrations of 200 pg/μL to 4000 ng/μL (depending on sample volume), providing a detection range of 4–4000 ng of DNA.

Common contaminants such as salts, free nucleotides, solvents, detergents, and protein are well tolerated in the dsDNA assays. The '1X' dsDNA assay kits provide reagent and buffer in a ready-to-use formulation and are highly selective for dsDNA over ssDNA, RNA, and protein.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay1 x dsDNA, high sensitivity
For Use With (Equipment)Microplate Reader
Product LineQuant-iT
Quantitation Range0.2 to 100 ng
Quantity1000 assays
Shipping ConditionWet Ice
Detection MethodFluorescence
FormatKit
Sample TypeDNA
Unit SizeEach

Frequently asked questions (FAQs)

What is the expiration date for the Quant-iT 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) (Cat. Nos. Q33232, Q3367)?

The expiration date for the Quant-iT 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) (Cat. Nos. Q33232, Q3367) is 6 months from the date of receipt.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

Citations & References (2)

Citations & References
Abstract
Total circulating cell-free miRNA in plasma as a predictive biomarker of the thyroid diseases.
Authors:Caglar O, Cayir A
Journal:J Cell Biochem
PubMed ID:30506793
Thyroid cancer is a common endocrine cancer. Great progress has been made in resolving its molecular mechanisms in recent years. The molecular changes observed in thyroid cancer can be used as biomarkers for diagnostic, prognostic and therapeutic purposes. MicroRNAs (miRNAs) are important components in biological and metabolic pathways, such as ... More
Tendon response to matrix unloading is determined by the patho-physiological niche.
Authors:Wunderli SL, Blache U, Beretta Piccoli A, Niederöst B, Holenstein CN, Passini FS, Silván U, Bundgaard L, Auf dem Keller U, Snedeker JG
Journal:Matrix Biol
PubMed ID:31917255
Although the molecular mechanisms behind tendon disease remain obscure, aberrant stromal matrix turnover and tissue hypervascularity are known hallmarks of advanced tendinopathy. We harness a tendon explant model to unwind complex cross-talk between the stromal and vascular tissue compartments. We identify the hypervascular tendon niche as a state-switch that gates ... More