For Clinicians
Resources for Medical Professionals  
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.  
  Food  
  Seasonal  
  Indoor  
  Allergic Asthma  
Is your patient a candidate for specific IgE testing?
Find out 
Get detailed information on whole allergens and allergen components. 
Allergen Encyclopedia 
Ready to test a patient?
Lab Ordering Guide 
Access videos and webinars delivered by key experts in the field of allergy.
On-Demand Education 
Frequently Asked Questions
About Allergy Testing
We're here to help
We get it. Diagnostic testing can seem confusing, intimidating, or even scary. That’s why we’ve put together some of our most asked questions to give you more information about specific IgE blood testing. Learn about the power of allergy diagnostic testing by exploring answers to some of the most common questions. It's time to take control of your allergy symptoms. 
What is an allergic sensitization test?
This is a blood test that is quick and simple. It measures the amount of allergen specific antibodies in the blood (IgE), which is an indicator of allergic sensitization. As a powerful diagnostic tool, it can test for hundreds of allergic triggers, such as pollen, mold, food, and animal dander and can be performed at any age. This blood test can help your healthcare professional determine if you are allergic, and to what.
What are the benefits of allergic sensitization testing?
There are many benefits to getting this blood test, also called a specific IgE (sIgE) blood test. Unlike a skin-prick test, an allergic sensitization test can be performed on anyone no matter the condition of their skin—even during an eczema flare-up. It can also be performed on someone while currently on medication, including antihistamines. This blood test is also safe to perform on someone who is pregnant. 
Is allergic sensitization testing for children or adults?
Anyone experiencing allergy-like symptoms can receive specific IgE (sIgE) blood testing. For babies and very young children, one blood sample collection is often less traumatic than the several scratches of a skin-prick test (SPT).
When should you test children for food allergies?
Sources such as the Canadian Pediatric Society discourage routine allergy testing prior to food introduction because positive results may be falsely interpreted as an allergy. Such a conclusion could lead to an unnecessary delay in allergenic food introduction—and might cause you to miss the important four-to-six-month window.1
The only exception may be infants at high risk of peanut allergy, i.e., those with severe eczema and/or egg allergy. In this case early tests such as a specific IgE blood test for peanut may be recommended.1  Again, talk with your pediatrician to determine the best course of action and learn more by reading our How to Introduce Food Allergens to Children blog.
How long does it take to get an allergy test?
Typically, it takes a few days to get blood test results back because the blood sample has to be sent to a lab for processing. 
How is an allergic sensitization test performed?
A small sample of blood is drawn and sent to a laboratory for analysis. The results are returned a few days later for your healthcare professional to interpret alongside your medical history and reported symptoms. This combination will help your healthcare professional confirm or rule out a suspected allergy.
If you don't see what's on your mind, check out our Facebook page or follow us on Instagram.
facebook   instagram
Tools for Understanding Allergies
Track allergy symptoms and prepare for a visit with a healthcare provider.
Learn about specific allergens, including common symptoms, management, and relief. 
Are you a healthcare provider? Get comprehensive information on hundreds of whole allergens and allergen components.
  1. Chan ES, Abrams EM, Hildebrand KJ, Watson W. Early introduction of foods to prevent food allergy. Allergy Asthma Clin Immunol. 2018 Sep 12;14(Suppl 2):57. doi: 10.1186/s13223-018-0286-1. PMID: 30275847; PMCID: PMC6157280. Available from: https://www.ncbi.nlm.nih.gov/pmc/articles/PMC6157280/.