Drug Allergy Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Adverse drug reactions (ADRs) are any harmful or unintended reaction to a drug that occurs at doses used for prevention, diagnosis, or treatment. They’re classified as either predictable reactions that may occur in anyone (Type A) or unpredictable reactions that occur in only susceptible individuals (Type B). Predictable reactions (Type A) (e.g., side effects, overdoses, and drug interactions) are the most common type of ADRs. Unpredictable reactions (Type B) occur in approximately 20 to 25 percent of patients who experience ADRs; these reactions are generally unrelated to the pharmacologic actions of the drug.1
A drug allergy, then, is one type of unpredictable (Type B) ADR, and it accounts for approximately 5 to 10 percent of all ADRs.1 To put it another way, a drug allergy is an abnormal immune-system reaction to a drug, be it prescription, over the counter, or herbal. Drug-allergy reactions can happen the first time the medication is taken or only after repeated exposure.2
Risk factors associated with developing a drug allergy include age (they’re more common in young/middle-aged adults), gender (they’re less common in men than women), presence of genetic polymorphisms and/or viral infections (e.g., HIV, herpes), previous drug reactions, and drug-related factors (e.g., the method of administration, single versus frequent dosing).1
Any drug can cause an allergic reaction, but some of the most common medications causing drug allergies include:1,2
Analgesics (aka pain killers) and antibiotics are the drug classes most commonly implicated in drug allergy and drug-induced anaphylaxis, but regional variations have been observed.4
Drug-allergy reactions can occur regardless of whether the drug is administered in liquid, pill, or injectable form.5 In addition, some drugs are cross-reactive.1 For example, in the case of allergy to β-lactam antibiotics, cross-reactions to penicillin, ampicillin, and amoxicillin are possible.3
Your healthcare provider may recommend a plan that includes the following.1,2,6
Your healthcare provider may direct you to take one of the following medications to improve your allergy symptoms:
If someone you’re with is having an allergic reaction and showing signs of shock, act quickly. Look for a weak, rapid pulse; trouble breathing; cool, clammy, and pale skin; confusion; and loss of consciousness. Immediately do the following:
Drug-allergy reactions most frequently and prominently affect the skin.1 In fact, allergic maculopapular rash, which often resembles measles or rubella, comprises an estimated 75 percent of all such reactions. However, responses also can manifest as organ or systemic reactions, such as serum sickness, various types of rash with eosinophilia and systemic symptoms (DRESS, drug rash with eosinophilia and systemic symptoms), and anaphylaxis.3
Symptoms of drug allergy can include:2
Drug allergy can also cause anaphylaxis, which can include symptoms such as:2
The following conditions and their related symptoms also can occur days or weeks after exposure and can persist after you stop taking the offending drug.2
Given the plethora of symptoms associated with drug allergy, diagnosis is particularly challenging.1
Together with your symptom history, specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Drug allergy can cause anaphylactic reactions, although this response is quite rare.3 That said, drugs account for 58.8 percent of all anaphylaxis-related death in the United States.4 Anaphylaxis is most commonly observed after administration of nonsteroidal anti-inflammatory agents, intravenous contrast agents, aspirin, and β-lactam antibiotics.3