For Clinicians
Resources for Medical Professionals  
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
  Food  
  Seasonal  
  Indoor  
  Allergic Asthma  
Is your patient a candidate for specific IgE testing?
Find out 
Get detailed information on whole allergens and allergen components. 
Allergen Encyclopedia 
Ready to test a patient?
On-Demand Education 
How do chemicals and pollution affect respiratory health?
July 2025  |  ✓  Medically Reviewed by: Gary Falcetano, PA-C, AE-C 
A girl experiencing allergies trying to decide if the cleaning products, perfume, cigarette smoke, laundry detergent , or other pollutants are causing her to have an allergic reaction.
An industrial chemical spill near your home is unlikely. However, you may encounter numerous chemicals in your daily life, such as when cleaning your home with over-the-counter products. What does it mean if you start sneezing and coughing while using them? Are these symptoms of a respiratory disease? Are these allergic reactions to chemicals?
Reactions triggered by chemicals and reactions triggered by allergens share many similar symptoms, but there is one important difference. When you have an allergic reaction, it’s your body's response to a normally harmless substance, such as pollen, mold, or pet dander. This overreaction stimulates the release of histamine and other substances that are responsible for common allergy symptoms like sneezing, coughing, runny nose, and itchy, watery eyes.1
Chemicals, on the other hand, don’t typically generate the production of histamine, which means they’re not true allergens. So, what is the role of chemicals in respiratory disease? And how can you find relief? Here, we answer these and other common questions about chemicals and allergies.
Can chemicals cause allergies?
Chemicals, on the other hand, don’t typically generate the production of histamine, which means they’re not true allergens. So, what is the role of chemicals in respiratory disease? And how can you find relief? Here, we answer these and other common questions about chemicals and allergies non-allergic rhinitis because your body isn’t releasing histamine as it would with an allergy. But irritation to your respiratory tract can increase the risk of these symptoms, and chemicals can be powerful irritants.2
Can pollution cause allergies or asthma?
The short answer is, it’s complicated. Allergies and asthma occur due to genetic and environmental factors, and the medical community is still researching how these factors interact. Air pollution itself is complex because it contains chemicals like ozone, nitrogen oxide, and nitrogen dioxide, as well as particles that we inhale. These particles, known as particulate matter, can penetrate deep into our lungs and even into our bloodstream. The effects of air pollution - even without long-term exposure - can lead to potential negative health effects. Exposure to fine particles comes from vehicle exhaust, industrial emissions, and smoke.3
Here are a few risks involved with exposure to and the effects of air pollution:
  • Some studies show that traffic-related pollution may cause asthma attacks and rhinitis in children.3,4
  • Other studies find that outdoor air pollution, like pollution from vehicle traffic, isn’t associated with allergy symptoms or diagnoses, but that poor air quality does cause irritation of the respiratory system.5
  • Exposure to air pollution (not just long-term exposure) may also be responsible for changing the way pollen, a common allergen, behaves in the environment. Pollutants can cause pollen to break into smaller pieces, and ozone in particular may cause changes to pollen grains’ structure, which can worsen their allergenic properties.3
All of this means that the impact of air pollution can potentially include allergic sensitivity or asthma. However, even if there isn’t a direct connection between pollutants and your particular symptoms, poor air quality, largely due to outdoor air pollution, remains a significant irritant that can cause symptoms similar to those of allergies and asthma.3
Will an air purifier help reduce air pollution?
Portable air purifiers can help remove indoor air pollution and improve indoor air quality. Upgrading the air filter in your heating and cooling system can also help. Either path will help reduce pollutants, but will not completely remove them.6
Am I allergic to cleaning chemicals?
If you’re scrubbing your floors or kitchen countertops and start sneezing, it’s not allergies. Cleaning chemicals such as bleach and detergent are a common cause of non-allergic rhinitis. This can result in allergy-like symptoms, but it’s your body reacting to an irritant, not producing an allergic reaction.
Can perfume or deodorant cause allergies?
No, your favorite perfume or cologne isn’t causing your significant other’s allergies or pulmonary disease. Still, they can certainly produce troubling allergy-like symptoms or make symptoms caused by allergic triggers, such as pollen or pet dander, be more intense. Body wash, deodorant, and other scented materials can be irritants, but they don’t cause an allergic reaction where the body releases histamine.7
Are chemicals making my allergies worse?
It’s possible, but it’s not as simple as it seems.  According to certain studies, chemical irritants and outdoor air pollution impact our airways and can aggravate allergies. Ozone, for example, has been found to increase sensitization to outdoor allergens.3 Pollutants may also interact with airborne allergic triggers like pollen, which can worsen allergy symptoms for those with sensitivities to those pollens. Overall, it’s recommended that anyone with a respiratory condition minimize their exposure to chemicals that may exacerbate their condition.8
How do I find relief for my symptoms?
The best way to address an allergy is to find out what specifically you’re allergic to. Your provider can order a specific IgE blood test, which, along with your history, helps determine which allergen or allergens may be causing your symptoms. With that information, you can avoid exposure to pollen, mold, or your particular triggers, and ensure you take any prescribed medication to manage your symptoms. If you’re experiencing symptoms around chemicals or you've been exposed to air pollution, and you know you have allergies, it’s especially important to control your symptoms and limit exposure to your triggers. 
If you don’t have a confirmed allergy but are experiencing sneezing, coughing, and other allergy-like symptoms, resist the urge to take allergy medication or antihistamines. Your body isn’t producing histamine so that these medications won’t work. In one study, 65% of patients taking a prescription antihistamine tested negative for an actual allergy, which means time and money were wasted on therapies that won’t bring the level of relief you need.9 (This is also why it’s a good idea to get an allergy test to help guide the selection of appropriate medications and to know what steps to take to reduce your exposure to your specific allergic triggers.)
What are the next steps?
If your symptoms persist around cleaning chemicals, perfumes, or air pollution, consult your healthcare provider about non-allergic rhinitis and potential triggers. They may be able to help you uncover the root cause of your symptoms and then guide you toward the best course of treatment for non-allergic rhinitis.
If you suspect your symptoms might be allergy-related, you can use our allergy symptom questionnaire to help track your symptoms, when they occur, how long they last, and the severity of the symptoms. You can use the results from the questionnaire to help inform a conversation with your healthcare provider.
By addressing concerns over air quality, chemicals, and pollutants in your environment, you can also take steps to improve your quality of life and overall respiratory health. 
Tools for Understanding Allergies
Track allergy symptoms and prepare for a visit with a healthcare provider.
Learn about specific allergens, including common symptoms, management, and relief. 
Are you a healthcare provider? Get comprehensive information on hundreds of whole allergens and allergen components.
  1. Histamine [Internet]. Cleveland Clinic; 2023 March 25 [cited 2025 May 5]. Available from: https://my.clevelandclinic.org/health/articles/24854-histamine.
  2. Wilbur S. Health Effects Classification and its Role in the Derivation of Minimal Risk Levels: Respiratory Effects. J Clean Technol Environ Toxicol Occup Med. 1998;7(3).
  3. González-Díaz SN, et al. Impact of air pollution in respiratory allergic diseases. Medicina Universitaria. 2016;18(73):212-215.
  4. Gehring U, et al. Traffic-related air pollution and the development of asthma and allergies during the first 8 years of life. Am J Respir Crit Care Med. 2010;181:596-603.
  5. Hirsch T, et al. Inner city air pollution and respiratory health and atopy in children. Eur Respir J. 1999;14(669-677).
  6. Air cleaners and air filters in the home [Internet]. US Environmental Protection Agency; 2025 March 11 [cited 2025 May 5]. Available from https://www.epa.gov/indoor-air-quality-iaq/air-cleaners-and-air-filters-home.
  7. What is a Sensitivity to Scents? [Internet]. Asthma and Allergy Foundation of America; 2022 May 18 [cited 2025 May 5]. Available from: https://community.aafa.org/db/ask-the-allergist/record/what-is-a-sensitivity-to-scents.
  8. Liccardi G, et al. Avoidance of allergens and air pollutants in respiratory allergy. Allergy. 2001;56:705-722.
  9. Szeinbach SL, et al. Identification of allergic disease among users of antihistamines. J Manag Care Pharm. 2004;10(3):234-238.