Optilite IgG subclass assays uniquely use a single international calibrant (ERM-DA470k) for both total IgG and subclasses, supporting:
Carvalho et al.1 reported:
"that the Immunoturbidimetry assay appears to be the most reliable to evaluate IgG Subclasses: the sum of IgG Subclasses and Total IgG correlate better than by Nephelometry."
Adapted from Carvalho et al. 20241
With over 40 years of protein diagnostics expertise, Binding Site’s IgG subclass assays are routinely used in more than 60 countries. Built on international reference alignment and proven assay performance, they deliver accurate, traceable results clinicians rely on.
Optilite assays combines scientific heritage with innovation — providing the accuracy and confidence laboratories need for every result.
The Optilite analyser delivers reliable performance and efficient workflows for every IgG subclass test. With onboard cooling, automated dilution management, long open vial stability and minimal operator intervention, laboratories can work with confidence and consistency.
Key benefits: