MGUS is of clinical significance
Monoclonal gammopathy of undetermined significance (MGUS) is characterised by the presence of a monoclonal protein in the serum of asymptomatic individuals who do not meet the diagnostic criteria for Multiple Myeloma, AL amyloidosis, Waldenström's macroglobulinaemia (WM), lymphoproliferative disorders, plasmacytoma or related conditions (learn more).
It was first described in 1978¹. The name was chosen as some patients with MGUS would go on to develop Multiple Myeloma but not all. Since then, many publications have demonstrated that MGUS is of clinical significance, even in patients who do not go on to develop Multiple Myeloma2.
MGUS is categorized into three types, depending on the type of monoclonal protein produced by the underlying clone. For all three types, the monoclonal protein must be present, at the concentrations presented in the table below, along with <10% clonal bone marrow cells, and an absence of the symptoms described in the bodily symptoms column3.
Diagnostic criteria |
||||||
% of MGUS patients |
Monoclonal protein |
Tumour cells |
Bodily symptoms |
Primary progression |
Progression rate per year |
|
IgG MGUS and IgA MGUS (referred to as non-IgM MGUS) |
66% | Serum IgG or IgA M-protein <30g/L |
<10% BMPC |
No CRAB* or amyloidosis | Multiple Myeloma, solitary plasmacytoma, immunoglobulin related amyloidosis | 1% |
IgM MGUS |
12% | Serum IgM M-protein <30g/L |
<10% Lympho-plasmactic infiltration |
No anemia, constitutional symptoms, hyperviscosity, lymphadenopathy, hepatosplenomegaly, or other end-organ damage |
Waldenström’s macroglobulinemia, Immunoglobulin related amyloidosis | 1.5% |
Light Chain Only MGUS |
19% | Abnormal κ/λ sFLC ratio Increased iFLC No Intact Ig on IFE |
<10% BMPC |
No CRAB or amyloidosis |
Light chain Multiple Myeloma, immunoglobulin light-chain amyloidosis | 0.3% |
*CRAB = hyperCalcemia, Renal impairment, Anemia and Bone lesions
Certain populations are at a higher risk of developing MGUS, such as older individuals, males, African Americans, and those with a family history of monoclonal gammopathy.
MGUS is frequently discovered as an incidental finding during laboratory tests that are conducted for other reasons9. This is because MGUS typically does not present with some specific symptoms that would lead to testing specifically for this condition.
The same initial testing is used no matter which monoclonal gammopathy is suspected. The initial testing aims to identify the presence of a monoclonal protein. Guidelines recommend use of sFLC testing plus SPE plus sIFE10.
Once a monoclonal protein is identified, the patient is further characterized to determine whether they have MGUS, SMM or MM (or another, rarer, monoclonal gammopathy). This includes tests for renal function, anemia, calcium levels, imaging to look for bone lesions and bone marrow biopsy to determine percentage of clonal bone marrow plasma cells3,11-13.
"A rise in serum free light chain concentration may precede an increase in intact immunoglobulin concentrations and may be the first sign that a patient is progressing from MGUS to Multiple Myeloma14."
Why is MGUS important?
As well as progression to Multiple Myeloma or another lymphoproliferative disorder, patients with MGUS may develop monoclonal protein-related disorders, these may include autoantibody activity by the monoclonal protein or deposition of the monoclonal protein in tissues and the associated organ dysfunction, or conditions associated with changes in the bone marrow microenvironment caused by the underlying tumour.
These changes can lead to increased infection, osteoporosis, fractures and thrombosis15. Patients with MGUS have an increased risk of death, not just through progression to Multiple Myeloma and other lymphoproliferative diseases but also bacterial infections, heart disease, liver diseases and kidney diseases2. This is why it is important to monitor patients with MGUS to detect these comorbidities and manage them.
Monitoring of patients with MGUS enables supportive care to be given if they develop any symptoms related to their monoclonal protein production, and also means that progression to Multiple Myeloma is detected earlier.
If progression to Multiple Myeloma is detected earlier, treatment can be given earlier, reducing the end organ damage the disease can cause, and improving patient outcome.
Patients with MGUS who are followed up and progress have fewer comorbidities at progression than patients first identified once they already have Multiple Myeloma15,16. They also have a longer median overall survival17.
Current guidelines recommend measuring monoclonal protein with serum free light chains (sFLC) and serum protein electrophoresis (SPE) at baseline, and then monitoring patients with SPE alone either annually or every two to three years, depending on the patient’s risk stratification at baseline18
However, these guidelines were last updated over 15 years ago. More recent data suggests that annual monitoring of all patients with MGUS using both sFLC and SPE may provide the best opportunity to identify patients progressing to multiple myeloma.
In the video below, listen to Prof. C. Ola Landgren, a member of the International Myeloma Working Group, discuss his recommendations for monitoring patients with MGUS.
Prof. Landgren was the lead author of a paper that presented evidence supporting annual testing, including sFLC and SPE, as a means to provide further information to clinicians and identify patients that were progressing to multiple myeloma.
A monoclonal gammopathy that does not reach the diagnostic criteria of an active malignancy such as Multiple Myeloma, AL amyloidosis or Waldenström’s macroglobulinemia but does affect organ function are called monoclonal gammopathy of clinical significance. The most common of these are monoclonal gammopathy of renal significance (MGRS)19-21 and monoclonal gammopathy of neurological significance (MGNS)22.
It is important for clinicians who manage patients with MGUS to be aware of the monoclonal gammopathies of clinical significance, so that symptoms can be identified if they occur, and treatment can be given if needed.