Search
Solve your chromatography separation challenges with our reversed phase (RP) HPLC and UHPLC columns.
Reversed phase HPLC and UHPLC columns offer a wide variety of stationary phases for the effective separation of your analytes. Reversed phase stationary phases are available in a range of surface chemistries that vary in the length and structure of their hydrocarbon chains. Our reversed phase column portfolio includes options for C18, C8, C4, C30, polar-embedded phases, and aromatic group phases like phenyl, biphenyl, phenyl-hexyl, and PFP. These stationary phases also come in different sorbents such as porous silica, solid core silica, and polymeric resin. By switching between different stationary phases, it is possible to control how strongly your molecules of interest are retained within your column.
Popular Thermo Scientific reversed phase HPLC and UHPLC column brands include:
Need to maximize your productivity and flexibility in method development? Thermo Scientific reversed phase HPLC columns are available in an array of chemistries to optimize separations and provide enhanced retention or changes in elution order. Reversed phase columns are one of the most popular modes in HPLC analysis.
Consistency you can rely on, results you can trust
Resolve impurities without feeling the pressure
Learn more about our small molecule HPLC and UHPLC column brands ›
| C18 (U)HPLC Columns | C8 (U)HPLC Columns | C4 (U)HPLC Columns |
![]() | ![]() | ![]() |
|
|
|
| Column | Column Advantages | Column Compatibility |
| Hypersil GOLD C18 Selectivity
|
| |
| Hypersil GOLD VANQUISH C18
|
| |
| Hypersil GOLD Peptide C18 |
| |
| Accucore C18
|
| |
| Accucore XL C18
|
| Uniguard Guard Cartridge Holders
|
| Accucore Vanquish C18+, 1.5 µm
|
|
| Column | Column Advantages | Column Compatibility |
| Hypersil GOLD C8
|
| Uniguard Guard Cartridge Holders
|
| Accucore C8
|
| Uniguard Guard Cartridge Holders
|
| Accucore XL C8
|
| Uniguard Guard Cartridge Holders
|
| Column | Column Advantages | Column Compatibility |
| Hypersil GOLD C4 |
| |
|
| Polar-Endcapped C18 (U)HPLC Columns | Amide-Embedded C18 (U)HPLC Columns |
![]() | ![]() |
|
|
| Column | Column Advantages | Column Compatibility |
| Hypersil GOLD aQ C18 Polar-Endcapped
|
| |
| Hypersil GOLD VANQUISH aQ
|
|
|
| Accucore aQ C18 Polar-Endcapped
|
| Uniguard Guard Cartridge Holders
|
| Column | Column Advantages | Column Compatibility |
| Accucore Polar Premium
|
| Uniguard Guard Cartridge Holders
|
| Accucore RP-MS (U)HPLC Columns | C30 (U)HPLC Columns |
| ![]() |
|
|
| Column | Column Advantage | Column Compatibility |
|
| Column | Column Advantage | Column Compatibility |
|
| Phenyl and Phenyl-Hexyl (U)HPLC Columns
| Biphenyl (U)HPLC Columns
| PFP (Pentafluorophenyl) (U)HPLC Columns
|
![]() | ![]() | ![]() |
|
|
|
| Column
| Column Advantages | Column Compatibility
|
| Uniguard Guard Cartridge Holders
| |
| Accucore Phenyl-Hexyl
|
| Uniguard Guard Cartridge Holders
|
| Column
| Column Advantages | Column Compatibility
|
| Accucore Biphenyl |
|
Column | Column Advantages | Column Compatibility |
| ||
| ||
|
|
|
|
EPA Method 543 - Quantitation of Organic Pesticides in Drinking Water Using Online Pre-concentration/Solid Phase Extraction and Tandem MS.
HRAM LC-MS method for the determination of nitrosamine impurities in drugs.
Fast and easy separation of 23 drugs of abuse including high, stable resolution of isobaric opioids from human urine by UHPLC-MS/MS.
Quantification of 25-hydroxyvitamin D2 and D3 in human plasma by tandem LC‑MS.
Hypersil GOLD HPLC columns - Outstanding peak shape for your separations.
This guide aides in troubleshooting of silica-based reversed phase columns.
* Required field
Need similar selectivity to C18 and C8 columns but less retention? C4 reversed phase HPLC columns provides outstanding sensitivity and high efficiency; and its shorter chain length and lower hydrophobicity make it useful for hydrophobic polypeptides and proteins.
Trying to separate aromatic isomers or halogenated compounds? PFP reversed phase HPLC columns offer alternative selectivity to C18 and are well suited to the selective analysis of halogenated and polar compounds containing hydroxyl, carboxyl, nitro, or other polar groups.
Need to retain polar compounds or use 100% aqueous mobile phase? These polar endcapped C18 reversed phase HPLC columns provide a controlled interaction mechanism by which polar analytes can be retained and exhibit superior stability towards highly aqueous mobile phase.