Search
There is a diverse range of protocols available in the literature around 3D cell culture and analysis. Discover those highlighted here to learn how researchers use Invitrogen and Gibco products in their methods.
| Cell type | Relevant growth factors | Relevant media | Key publication |
|---|---|---|---|
| Pluripotent stem cells | Activin, FGF4, FGF2, Noggin, SHH | RPMI1640, Advanced DMEM/F12 | Dye, BR., et al., Elife. 2015 |
| Pluripotent stem cells | FGF2 | Advanced DMEM/F12 | Wilkinson, DC., et al., Stem Cells Translational Medicine. 2016 |
| Cell type | Relevant growth factors | Relevant media | Key publication |
|---|---|---|---|
| Pluripotent stem cells | Activin, FGF4, FGF2, Noggin, SHH | RPMI1640, Advanced DMEM/F12 | Dye, BR., et al., Elife. 2015 |
| Pluripotent stem cells | FGF2 | Advanced DMEM/F12 | Wilkinson, DC., et al., Stem Cells Translational Medicine. 2016 |
To help guide you in your 3D cell culture, our R&D teams have developed suggested protocols for the generation of spheroids from the most commonly used cancer cell lines using Nunclon Sphera cell culture plastics and Gibco media.
Cancer organoids are known to have complex culture requirements that can be challenging. The OncoPro Tumoroid Culture Medium Kit helps simplify tumoroid culture with well-defined, easy-to-follow protocols, helping make physiologically relevant cancer models more accessible.
For Research Use Only. Not for use in diagnostic procedures.