TOPO PCR Cloning
Twenty years ago, the TOPO name became synonymous with superior quality and game-changing innovation in PCR cloning. Today, Invitrogen TOPO cloning kits are still the fastest, simplest, and most efficient cloning solutions available. There is no substitute.
TOPO PCR Cloning Selection Tool
TOPO PCR Cloning Handbook
TOPO cloning technology highlights
  • Fast—5-minute, room temperature reaction
  • Simple—add restriction sites and/or universal primer sites to either end of your PCR product in just 3 easy steps
  • Efficient—up to 95% of clones contain desired insert
  • Flexible—available in a variety of formats and sizes to suit your choice in polymerase
  • Proven—20,000 citations
No matter what your downstream application is, TOPO PCR cloning solutions help you get the right clone sooner, freeing up your time to answer more important questions.
TOPO Tip
Eliminate blue/white screening with TOPO Zero Blunt vectors, which contain a lethal ccdB gene. When disrupted by the insertion of a PCR product, only positive recombinants are permitted to grow upon transformation.
Choose the right TOPO cloning solution for you
Invitrogen TOPO PCR cloning technology was developed to help improve cloning efficiency, simplify protocol setup, and accommodate a wide range of PCR insert sizes. TOPO technology enables inserts with compatible ends to be readily joined to the vector in 5 minutes, without the need for additional ligation steps. Learn more about our TOPO cloning solutions below.
For the direct insertion of blunt-end PCR products amplified with proofreading thermostable polymerases into a plasmid vector. Now with 25% extra reactions.
For the direct insertion of Taq polymerase–amplified PCR products into a plasmid vector for subcloning. Now with 25% extra reactions.
For easy, highly efficient cloning of extra-long PCR products (up to 13kb) using the new linearized, topoisomerase I-activated pCR™-XL-2-TOPO vector.
TOPO Kits without competent cells
Gateway recombinant cloning with TOPO Technology
Expression cloning with TOPO Technology
Featured video
See how TOPO PCR Cloning technology works
Learn how simple it is to get 95% recovery of your clone in just 5 minutes.
Meet the Inventor
Find out how the revolutionary TOPO cloning was developed with Dr. Jon Chestnut, research fellow in Synthetic Biology R&D.
Dr. Jon Chesnut
TOPO Cloning Technology
Cloning Support Center
Find tips, troubleshooting help, and resources for your cloning applications
Cloning Education
Learn about PCR cloning strategies and subcloning approaches
PCR Education
Review the principles of PCR, components, cycling parameters, methods, and applications