Search
Search

| Assay Name: | rno-miR-186-3p |
|---|---|
| miRBase Accession Number: | MI0000931 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCCCAAAGGUGAAUUUUUUGG |
| Species: | Rat, Cricetulus griseus |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | GCCCAAAGGUGAAUUUUUUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-186-3p | MIMAT0017143 |
| Cricetulus griseus | cgr-miR-186-3p | MIMAT0023814 |