Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12818
          Assay Name: rno-miR-532-5p
          miRBase Accession Number: MI0006154
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 16109870 - 16109948 [+] on Build Rnor_6.0
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Species: Rat, Dasypus novemcinctus, Otolemur garnettii, Pteropus alecto, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC12818

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000113, mir-188

          Mature miRNA Sequence:

          CAUGCCUUGAGUGUAGGACUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-532-5p MIMAT0005322
          Dasypus novemcinctus dno-miR-532-5p MIMAT0047963
          Otolemur garnettii oga-miR-532 MIMAT0049692
          Pteropus alecto pal-miR-532-5p MIMAT0039901
          Rabbit ocu-miR-532-5p MIMAT0048519

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-532
          Stem-loop Accession # MI0006154
          Stem-loop Sequence
          UUUUCUCUUCCAUGCCUUGAGUGUAGGACUGUUGACAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGAGCAAG
          Chromosome Location Chr. X - 16109870 - 16109948 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005322
          miRBase ID: rno-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Chromosome Location: Chr. X - 16109870 - 16109948 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-532
          Stem-loop Accession # MI0039148
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUGGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047963
          miRBase ID: dno-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-532
          Stem-loop Accession # MI0040476
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUUGACAUCUUAAUUACCCUCCCACACCCAAGGCUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049692
          miRBase ID: oga-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-532
          Stem-loop Accession # MI0032391
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUUGCCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039901
          miRBase ID: pal-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-532
          Stem-loop Accession # MI0039442
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUUGGAAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048519
          miRBase ID: ocu-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465601_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12818
          Ambion® Pre-miR™ miRNA Precursor : PM12818
          TaqMan™ MicroRNA Assay : 002051
          mirVana® miRNA inhibitor : MH12818
          TaqMan™ Advanced miRNA Assay : rno481405_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0