Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12949
          Assay Name: hsa-miR-518e-5p
          miRBase Accession Number: MI0003169
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53729838 - 53729925 [+] on Build GRCh38
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Species: Human, Gorilla gorilla, Pan troglodytes, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12949

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000020, mir-515

          Mature miRNA Sequence:

          CUCUAGAGGGAAGCGCUUUCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-518e-5p MIMAT0005450
          Human hsa-miR-519a-5p MIMAT0005452
          Human hsa-miR-519b-5p MIMAT0005454
          Human hsa-miR-519c-5p MIMAT0002831
          Human hsa-miR-522-5p MIMAT0005451
          Human hsa-miR-523-5p MIMAT0005449
          View More Rows
          Gorilla gorilla ggo-miR-518e-5p MIMAT0036427
          Gorilla gorilla ggo-miR-519a-5p MIMAT0036439
          Gorilla gorilla ggo-miR-519c-5p MIMAT0036410
          Gorilla gorilla ggo-miR-519c MIMAT0036435
          Gorilla gorilla ggo-miR-522-5p MIMAT0036437
          Gorilla gorilla ggo-miR-523-5p MIMAT0036414
          Pan troglodytes ptr-miR-519f-5p MIMAT0036456
          Rhesus monkey mml-miR-519a MIMAT0036445

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-518e
          Stem-loop Accession # MI0003169
          Stem-loop Sequence
          UCUCAGGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGGCUAAAAGAAAAGAAAGCGCUUCCCUUCAGAGUGUUAACGCUUUGAGA
          Chromosome Location Chr. 19 - 53729838 - 53729925 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005450
          miRBase ID: hsa-miR-518e-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53729838 - 53729925 [+] on Build GRCh38

          Stem-loop ID hsa-mir-519a-1
          Stem-loop Accession # MI0003178
          Stem-loop Sequence
          CUCAGGCUGUGACACUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAGGAAAGUGCAUCCUUUUAGAGUGUUACUGUUUGAG
          Chromosome Location Chr. 19 - 53752397 - 53752481 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005452
          miRBase ID: hsa-miR-519a-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53752397 - 53752481 [+] on Build GRCh38

          Stem-loop ID hsa-mir-519b
          Stem-loop Accession # MI0003151
          Stem-loop Sequence
          CAUGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAGUGCAUCCUUUUAGAGGUUUACUGUUUG
          Chromosome Location Chr. 19 - 53695213 - 53695293 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005454
          miRBase ID: hsa-miR-519b-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53695213 - 53695293 [+] on Build GRCh38

          Stem-loop ID hsa-mir-519c
          Stem-loop Accession # MI0003148
          Stem-loop Sequence
          UCUCAGCCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAGUGCAUCUUUUUAGAGGAUUACAGUUUGAGA
          Chromosome Location Chr. 19 - 53686469 - 53686555 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002831
          miRBase ID: hsa-miR-519c-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53686469 - 53686555 [+] on Build GRCh38

          Stem-loop ID hsa-mir-522
          Stem-loop Accession # MI0003177
          Stem-loop Sequence
          UCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGA
          Chromosome Location Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005451
          miRBase ID: hsa-miR-522-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38

          Stem-loop ID hsa-mir-523
          Stem-loop Accession # MI0003153
          Stem-loop Sequence
          UCUCAUGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAACGCGCUUCCCUAUAGAGGGUUACCCUUUGAGA
          Chromosome Location Chr. 19 - 53698385 - 53698471 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005449
          miRBase ID: hsa-miR-523-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: Chr. 19 - 53698385 - 53698471 [+] on Build GRCh38

          Gorilla gorilla

          Stem-loop ID ggo-mir-518e
          Stem-loop Accession # MI0031082
          Stem-loop Sequence
          CAAGAUCUCAGGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGGCUAAAAGAAAAGAAAGCGCUUCCCUUCAGAGUGUUACGCUUUGAGAAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036427
          miRBase ID: ggo-miR-518e-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-519a-1
          Stem-loop Accession # MI0031090
          Stem-loop Sequence
          GCAAGAAGAUCUCAGGCUGUGACACUCUAGAGGGAAGCGCUUUCUGUUUUCUGAAAAAGAAAGUGCAUCCUUUUAGAGUGUUACUGUUUGAGAAAAGCAACGUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036439
          miRBase ID: ggo-miR-519a-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-519c-1
          Stem-loop Accession # MI0031071
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCCCAUGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAGAGAAAGUGCAUCCUUUUAGAGGUUUACUGUUUGAGGAAAGCAGCGGUGAAGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036410
          miRBase ID: ggo-miR-519c-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-519c-2
          Stem-loop Accession # MI0031087
          Stem-loop Sequence
          GCAAGAAGAUCUCAGCCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAGGAAAGUGCUUUCUUUCAGAGGGUUACGGUUUGAGAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036435
          miRBase ID: ggo-miR-519c
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-522
          Stem-loop Accession # MI0031089
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGAAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036437
          miRBase ID: ggo-miR-522-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-523
          Stem-loop Accession # MI0031073
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCUCAUGCUGUGACCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAACGCGCUUCCCUAUAGAGGGUUACCCUUUGAGAAAAGCAGCAUUGAAGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036414
          miRBase ID: ggo-miR-523-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-519f
          Stem-loop Accession # MI0031110
          Stem-loop Sequence
          GCAAGAAGACCUCAGGCUGUGACACUCUAGAGGGAAGCGCUUUCUGUUUUCUGAAAAAGAAAGUGCAUCCUUUUAGAGUGUUACUGUUUGAGAAAAGCAACGUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036456
          miRBase ID: ptr-miR-519f-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-519a-2
          Stem-loop Accession # MI0031097
          Stem-loop Sequence
          GUGCUGGAGCAAGAAGAUCUCAGGCUCUAACCCUCUAGAGGGAAGCGCUUUCUGUCGUCUGAGAGAGAAGAAAGUGCAUCCUUUUAGAGGAUUACGGUUUGAGAAAAGCAAUGGUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036445
          miRBase ID: mml-miR-519a
          Mature miRNA Sequence: CUCUAGAGGGAAGCGCUUUCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303957_pri
          TaqMan™ Pri-miRNA Assay : Hs03295478_pri
          TaqMan™ Pri-miRNA Assay : Hs03295797_pri
          TaqMan™ Pri-miRNA Assay : Hs03296239_pri
          TaqMan™ Pri-miRNA Assay : Hs04225384_pri
          TaqMan™ Pri-miRNA Assay : Hs03304060_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12949
          Ambion® Pre-miR™ miRNA Precursor : PM12949
          TaqMan™ MicroRNA Assay : 002371
          mirVana® miRNA inhibitor : MH12949
          TaqMan™ Advanced miRNA Assay : 479491_mir


          miRBase Alias:

          Human: hsa-miR-518e* (v17)
          Human: hsa-miR-522* (v17)
          Human: hsa-miR-523* (v17)
          Human: hsa-miR-519a* (v17)
          Pongo pygmaeus: ppy-miR-519c-5p (v21)
          Pongo pygmaeus: ppy-miR-519b-5p (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline