Search
Search

| Assay Name: | mmu-miR-486b-3p |
|---|---|
| miRBase Accession Number: | MI0014103 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCAGCUCAGUACAGGA |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | CGGGGCAGCUCAGUACAGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-486b-3p | MIMAT0014944 |